Would you like to log in again?
IntelliCAL™ Sodium ISE Analysis Package, Hach
Supplier: Hach
ISENA381AP ISENA381AP
89501-110EA 1272.87 USD InStock
CA89501-110 89501-110
IntelliCAL™ Sodium ISE Analysis Package, Hach
Electrodes ISE Electrodes
The IntelliCAL™ sodium ISE analysis package contains the IntelliCAL™ ISENA381 Sodium ISE, 100 pack of sodium ISA powder pillows, 1 L of 100 mg/L sodium standard solution, 500 ml of 1000 mg/L sodium standard solution, 0.02 M ammonium chloride electrode filling solution, fill solution dispensing kit.
This package provides fast, stable, and accurate responses, and it has ultimate traceability in measurement history. The user is alerted when re-calibration is needed, and componants can be moved between meters without the need to re-calibrate or re-enter measurement settings. Thermistor is integrated.
IntelliCAL™ ISENa381 is a digital, combination sodium ion selective electrode (ISE) with a refillable double junction reference and built-in temperature sensor. The IntelliCAL ISENa381 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENa381 is ideal for measuring sodium concentrations in wastewater, drinking water and general water quality applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty