Would you like to log in again?
Standard Shoulder Split Leather Palm Gloves, Wells Lamont
Supplier: Wells Lamont
Y3201L Y3201M
80092-098EA 7.72 USD InStock
80092-098 32935-120
Standard Shoulder Split Leather Palm Gloves, Wells Lamont
Gloves
Wells Lamont Industrial’s Y3201 shoulder split leather palm glove is a great choice for an everyday work glove.
  • Standard shoulder split cowhide leather palm
  • Striped canvas back and cuff
  • Full leather palm, fingertips and knuckle strap
  • Wing thumb and safety cuff
  • Lined palm
This durable glove offers split leather index fingers, leather tips, and a leather knuckle strap. The lined palm provides added comfort in the palm of the hand.
The safety cuff coverage protects just over the wrist area for added protection and is easy to take off and on. The wing thumb provides a good fit to perform any task.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty