Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Diphenylphosphino)biphenyl 97%

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Warning

H61640-06 H61640-14 H61640-14 H61640-06
AAH61640-14EA 372.59 USD InStock
AAH61640-14 CAAAH61640-06 AAH61640-06 CAAAH61640-14
2-(Diphenylphosphino)biphenyl 97%
2-(Diphenylphosphino)biphenyl


Formula: C24H19P
MW: 338.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11559063
CAS Number: 13885-09-1
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty