Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Somatostatin 14

Supplier: Thermo Scientific
J66168-LB0
AAJ66168-LB0EA 216.96 USD InStock
AAJ66168-LB0
Somatostatin 14
Proteins and Peptides
Somatostatin 14

CAS: 38916-34-6
Form: Powder
MDL: MFCD00076762

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty