Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Parathyroid Hormone (1-34)

Supplier: Thermo Scientific
J66180-MCR J66180-EXD
AAJ66180-EXDEA 218.71 USD InStock
AAJ66180-EXD AAJ66180-MCR
Human Parathyroid Hormone (1-34)
Proteins and Peptides
Parathyroid Hormone (1-34), human

CAS: 52232-67-4
Form: Solid
MDL: MFCD00149013

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty