Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Influenza Matrix Peptide (58-66)

Supplier: Thermo Scientific
J66181-MCR J66181-EXD
AAJ66181-MCREA 242.28 USD InStock
AAJ66181-MCR AAJ66181-EXD
Influenza Matrix Peptide (58-66)
Proteins and Peptides
Influenza Matrix Peptide (FluM1 58-66)

CAS: 141368-69-6
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty