Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agarose, powder, Genetic Technology Grade

Supplier: Thermo Scientific Chemicals

J66208-14 J66208-22
AAJ66208-14EA 152.17 USD InStock
AAJ66208-14 AAJ66208-22
Agarose, powder, Genetic Technology Grade
Agarose
Agarose, Genetic Technology Grade

Applications: Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes


Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty