Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glucagon like peptide 1 (7-37)

Supplier: Thermo Scientific
J66232-MCR J66232-EXD
AAJ66232-EXDEA 199.86 USD InStock
AAJ66232-EXD AAJ66232-MCR
Glucagon like peptide 1 (7-37)
Proteins and Peptides
Glucagon Like Peptide-1 (7-37)

CAS: 106612-94-6
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty