Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Somatostatin 28 (1-14)

Supplier: Thermo Scientific
J66261-MCR
AAJ66261-MCREA 206.97 USD InStock
AAJ66261-MCR
Somatostatin 28 (1-14)
Proteins and Peptides
Somatostatin 28 (1-14)

CAS: 79243-10-0
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty