Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Somatostatin 28

Supplier: Thermo Scientific
J66274-MCR J66274-EXD
AAJ66274-EXDEA 285.43 USD InStock
AAJ66274-EXD AAJ66274-MCR
Somatostatin 28
Proteins and Peptides
Somatostatin 28

CAS: 73032-94-7
Form: Solid
MDL: MFCD00076772

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty