Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Neuropeptide Y (3-36)

Supplier: Thermo Scientific
J66381-EXD J66381-MCR
AAJ66381-MCREA 239.43 USD InStock
AAJ66381-MCR AAJ66381-EXD
Human Neuropeptide Y (3-36)
Proteins and Peptides
Neuropeptide Y (3-36), human

CAS: 150138-78-6
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty