Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat [Tyr8] Atrial Natriuretic Peptide (5-27)

Supplier: Thermo Scientific
J66452-MA J66452-LB0
AAJ66452-LB0EA 715.19 USD InStock
AAJ66452-LB0 AAJ66452-MA
Rat [Tyr8] Atrial Natriuretic Peptide (5-27)
Proteins and Peptides
[Tyr8] Atrial Natriuretic Peptide (5-27), rat

Molecular Formula: C98H156N34O33S2
Formula Weight: 2402.62
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00076236

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty