Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agarose, powder for electrophoresis

Supplier: Thermo Scientific Chemicals

J66501-18 J66501-30
AAJ66501-18EA 124.97 USD InStock
AAJ66501-18 AAJ66501-30
Agarose, powder for electrophoresis
Agarose
Agarose, Electrophoresis Grade

Applications: Standard agarose suitable for most general applications. Good gel strength and clarity


Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty