Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Arg^8] Vasotocin

Supplier: Thermo Scientific
J66551-MCR
AAJ66551-MCREA 82.2 USD InStock
AAJ66551-MCR
[Arg^8] Vasotocin
Proteins and Peptides
[Arg^8] Vasotocin

CAS: 74927-14-3
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty