Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse;Rat Myelin Oligodendrocyte Glycoprotein Peptide (35-55)

Supplier: Thermo Scientific
J66557-MCR J66557-EXD
AAJ66557-MCREA 378.37 USD InStock
AAJ66557-MCR AAJ66557-EXD
Mouse;Rat Myelin Oligodendrocyte Glycoprotein Peptide (35-55)
Proteins and Peptides
Myelin Oligodendrocyte Glycoprotein Peptide (35-55), rat, mouse

CAS: 163913-87-9
Form: Powder
MDL: MFCD02265607

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty