Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

beta-Amyloid (1-16)

Supplier: Thermo Scientific
J66562-MA J66562-LB0
AAJ66562-LB0EA 422.13 USD InStock
AAJ66562-LB0 AAJ66562-MA
beta-Amyloid (1-16)
Proteins and Peptides
Amyloid ^b (1-16)

CAS No.: 131580-10-4
Molecular Formula: C84H119N27O28
Formula Weight: 1955.00
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Solubility: Soluble in water or aqueous buffer
MDL No.: MFCD00214592

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty