Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Lys^8] Vasopressin

Supplier: Thermo Scientific
J66728-MCR
AAJ66728-MCREA 66.64 USD InStock
AAJ66728-MCR
[Lys^8] Vasopressin
Proteins and Peptides
[Lys^8] Vasopressin

CAS: 50-57-7
Form: Solid
MDL: MFCD00133970

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty