Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Neuropeptide Y

Supplier: Thermo Scientific
J66797-MCR J66797-EXD
AAJ66797-EXDEA 229.25 USD InStock
AAJ66797-EXD AAJ66797-MCR
Human Neuropeptide Y
Proteins and Peptides
Neuropeptide Y, human

CAS: 90880-35-6
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty