Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat Calcitonin Gene Related Peptide

Supplier: Thermo Scientific
J66827-MCR J66827-EXD
AAJ66827-MCREA 618.93 USD InStock
AAJ66827-MCR AAJ66827-EXD
Rat Calcitonin Gene Related Peptide
Proteins and Peptides
Calcitonin Gene Related Peptide, rat

CAS: 96827-03-1
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty