Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human;Rat Corticotropin-Releasing Factor

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

J66835-EXD
AAJ66835-EXDEA 250.54 USD InStock
AAJ66835-EXD
Human;Rat Corticotropin-Releasing Factor
Proteins and Peptides
Corticotropin-Releasing Factor, human, rat

CAS: 86784-80-7
Form: Solid
MDL: MFCD00213806

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty