Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fibronectin-Binding Protein

Supplier: Thermo Scientific
J66903-EXD J66903-MCR
AAJ66903-EXDEA 188.69 USD InStock
AAJ66903-EXD AAJ66903-MCR
Fibronectin-Binding Protein
Proteins and Peptides
Fibronectin-Binding Protein

CAS: 119977-20-7
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty