Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Endomorphin-2

Supplier: Thermo Scientific
J66953-MB J66953-LB0
AAJ66953-MBEA 445.39 USD InStock
AAJ66953-MB AAJ66953-LB0
Endomorphin-2
Proteins and Peptides
Endomorphin-2

CAS No.: 141801-26-5
Molecular Formula: C32H37N5O5
Formula Weight: 571.67
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD01321064

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty