Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human;Pig [Leu^1^3] Motilin

Supplier: Thermo Scientific
J66962-MCR J66962-EXD
AAJ66962-EXDEA 193.73 USD InStock
AAJ66962-EXD AAJ66962-MCR
Human;Pig [Leu^1^3] Motilin
Proteins and Peptides
[Leu^1^3] Motilin, human, porcine

CAS: 116283-54-6
Form: Solid
MDL: MFCD00133964

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty