Would you like to log in again?
IntelliCAL™ Chloride ISE Analysis Package, Hach
Supplier: Hach
ISECL181AP ISECL181AP
89501-102EA 1660.66 USD InStock
CA89501-102 89501-102
IntelliCAL™ Chloride ISE Analysis Package, Hach
Electrodes ISE Electrodes
The IntelliCAL™ Chloride ISE Analysis package contains the IntelliCAL™ ISECL181 Chloride ISE, 100 pack of chloride ISA powder pillows, 1 L of 100 mg/L chloride standard solution, 500 ml of 1000 mg/L chloride standard solution.
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
IntelliCAL™ ISECL181 is a digital, combination chloride ion selective electrode (ISE) with a non-refillable, double junction reference and built-in temperature sensor. The IntelliCAL ISECL181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISECL181 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications. The ISECL181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
Dry storage and fast response time. No replacement membranes. Integrated thermistor. Solid-state crystal membrane sensor type.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty