Would you like to log in again?
IntelliCAL™ Nitrate ISE Analysis Package
Supplier: Hach
ISENO3181AP
89501-122EA 2313.71 USD InStock
89501-122
IntelliCAL™ Nitrate ISE Analysis Package
Electrodes ISE Electrodes
IntelliCAL™ ISENO3181 is a digital, combination nitrate ion selective electrode (ISE) with a non-refillable, double junction reference and built-in temperature sensor.
  • Utilizes a unique solid-state sensor technology that eliminates costly membrane replacements
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Special features include dry storage and fast response time, and no replacement membranes. Integrated thermistor.
The IntelliCAL ISENO3181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENO3181 is ideal for measuring nitrate concentrations in wastewater, drinking water and general water quality applications. The ISENO3181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
Ordering information: The IntelliCAL™ Nitrate ISE Analysis Package contains the IntelliCAL ISENO318101 Nitrate ISE, 100 pack of nitrate ISA powder pillows, 500 ml of 1.0 mg/L nitrogen-nitrate standard solution, 500 ml of 10 mg/L nitrogen-nitrate standard solution, 500 ml of 100 mg/L nitrogen-nitrate standard solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty