Would you like to log in again?
HQ430d Benchtop Meter Package with CDC401 Conductivity Cell, Hach
Supplier: Hach
8509600 8509600
89500-880EA 4920.43 USD InStock
89500-880 CA89500-880
HQ430d Benchtop Meter Package with CDC401 Conductivity Cell, Hach
Electrochemical Meters Conductivity Meters
The Hach HQ430d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • Flexible benchtop meter makes your lab testing of critical electrochemistry parameters more efficient
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements - IntelliCAL™ smart probes store all calibrations in the probe
  • Enhance productivity with an ultra-bright screen and large font size for easy to read results
  • Convenient kit includes everything you need to start testing
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous lab use. HQd meters feature a simple user interface that does not require manuals or training to operate. The HQ430d is a flexible multi-parameter meter that can expand to addtional parameters as testing needs change. Included in this package is a standard, low-maintenance conductivity probe suitable for general purpose measurement.
Ordering information: Includes HQ430d Benchtop Meter, standard CDC401 conductivity/TDS/salinity cell with 1 m cable, NaCl conductivity standard (1000 S/cm, 100 mL), probe stand, 4 AA batteries, power adapter, USB cable for data transfer, quick-start guide, user manual, and documentation CD.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty