Would you like to log in again?
HQ440d Benchtop Meter Package for Pure Water Applications with PHC281 pH Ultra Electrode and CDC401 Conductivity Cell, Hach
Supplier: Hach
8510200
89500-882EA 7462.02 USD InStock
89500-882
HQ440d Benchtop Meter Package for Pure Water Applications with PHC281 pH Ultra Electrode and CDC401 Conductivity Cell, Hach
Electrochemical Meters pH and Multi-Parameter Meters
The Hach HQ440d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • Flexible benchtop meter makes your lab testing of critical electrochemistry parameters more efficient
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements - IntelliCAL™ smart probes store all calibrations in the probe
  • Enhance productivity with an ultra-bright screen and large font size for easy to read results
  • Convenient kit includes everything you need to start testing
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous lab use. HQd meters feature a simple user interface that does not require manuals or training to operate. The high-flowing reference and open junction of the pH Ultra electrode included in this package delivers fast stablilzation time even in the most difficult sample types and low ionic strength waters.
Ordering information: Includes HQ440d Benchtop Meter, PHC281 pH Ultra electrode with 1 m cable, CDC401 conductivity/TDS/salinity cell with 1 m cable, pH Buffer Kit (500 mL), pH storage solution (500 mL), NaCl conductivity standard (1000 S/cm, 100 mL), probe stand, 4 AA batteries, power adapter, USB cable for data transfer, quick-start guide, user manual, and documentation CD.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty