Would you like to log in again?
POWERSTAT, 0 to 140 V, Ace Glass Incorporated
Supplier: Ace Glass
12084-20
89076-328EA 1766.18 USD InStock
89076-328
POWERSTAT, 0 to 140 V, Ace Glass Incorporated
Transformers
This variable transformer is ideally suited for use in applications requiring a portable source of variable A/C voltage up to 1.4KVA capacity
Controls on panel are recessed for eye appeal and protection from accidental bumping. Equipped with grounded NEMA cord-plug assembly, on-off switch, pilot light, output receptacle and fuse. Supplied with slots at rear that accept wall hanger brackets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty