Would you like to log in again?
Flourescent Mounting Media, KPL
Supplier: KPL
5570-0005
95059-434EA 103.35 USD InStock
95059-434
Flourescent Mounting Media, KPL
Stains Microscopy and staining reagents
Fluorescent Mounting Media is glycerol-based and optically clear liquid for immunofluorescence microscopy
It is formulated to retard photobleaching of fluorochromes such as fluorescein and rhodamine. The mounting media prevents photobleaching and fading of floiurochromes. Can be used as a dilutent for flourescent conjugates. It is not a permanent mounting medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty