Would you like to log in again?
Heat Exchanger Coil Elbow Adapter, Ace Glass Incorporated
Supplier: Ace Glass
12067-06 12067-04 12067-02
89076-228EA 221.64 USD InStock
89076-228 89076-226 89076-224
Heat Exchanger Coil Elbow Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These are stainless steel elbow adapters for connecting a heat exchange coil tubing to a female pipe thread (FNPT).
  • Stainless steel
  • Makes a compression seal
  • Working pressure of 25 psig
Adapter fits over tubing and makes a compression seal. Working pressure of 25 psig. Complete item includes compression adapter, elbow, and pipe nipple.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty