Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) carbonate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Lead carbonat

1.07381.0250 1.07381.0250
EM1.07381.0250CS 10876.52 USD InStock
CA1.07381.0250 EM1.07381.0250
Lead(II) carbonate, EMSURE® ACS for analysis, Supelco®
Lead (II) carbonate


Formula: PbCO₃
MW: 267.21 g/mol
Melting Pt: 315 °C
Density: 6.43 g/cm³ (20 °C)
MDL Number: MFCD00064631
CAS Number: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Melting Point 315 °C (decomposition)
Molar Mass 267.21 g/mol
Bulk Density 1300 kg/m3
Density 6.6 g/cm3 (20 °C)
pH 5.8 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 6.1
Hazard Class Description Toxic materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty