Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 40% for determination of gas metabolism acc. to Knipping, Supelco®

Supplier: MilliporeSigma
Danger

1.09286.2500 1.09286.2500
EM1.09286.2500CS 501 USD InStock
EM1.09286.2500 CA1.09286.2500
Sulfuric acid 40% for determination of gas metabolism acc. to Knipping, Supelco®
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) soluble,(caution ! development of heat)
Melting Point -68 °C
Boiling Point 119 °C
Density 1.30 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty