Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neodymium(III) oxide ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Dineodymium trioxideNeodymium oxide

M-Clarity™ quality level = MQ200

1.12276.0025 1.12276.0025
EM1.12276.0025CS 3354 USD InStock
CA1.12276.0025 EM1.12276.0025
Neodymium(III) oxide ≥99%, Sigma-Aldrich®
Neodymium (III) oxide


Formula: Nd₂O₃
MW: 336.48 g/mol
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2270 °C
Density: 7.24 g/cm³ (20 °C)
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Melting Point 2272 °C
Molar Mass 336.48 g/mol
Bulk Density 1200 kg/m3
Density 7.24 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty