Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Samarium(III) oxide ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

1.12321.0010 1.12321.0010
EM1.12321.0010CS 27621 USD InStock
CA1.12321.0010 EM1.12321.0010
Samarium(III) oxide ≥99%, Sigma-Aldrich®
Samarium (III) oxide


Formula: O₃Sm₂
MW: 348.72 g/mol
Melting Pt: 2325 °C
Density: 7.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011237
CAS Number: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,08347
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 2325 °C
Molar Mass 348.72 g/mol
Bulk Density 850 kg/m3
Density 8.347 g/cm3
pH 8.0 (10 g/l, H2O, 25 °C) (slurry)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty