Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bismuth ≥99%, powder, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

1.12400.0100 1.12400.0100
EM1.12400.0100CS 11665 USD InStock
CA1.12400.0100 EM1.12400.0100
Bismuth ≥99%, powder, Sigma-Aldrich®
Bismuth


Formula: Bi
MW: 208.98 g/mol
Density: 9.78 g/cm³ (20 °C)
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
ADR: 4.1,III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 271 °C
Molar Mass 208.98 g/mol
Bulk Density 6000 kg/m3
Boiling Point 1564 °C (1013 hPa)
Vapor Pressure 1 hPa (840 °C)
Density 9.78 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty