Would you like to log in again?
Extrelut Nt Refill Pack For Column Fillings Extrelut Nt 20
Supplier: MilliporeSigma
1.15093.0001 1.15093.0001
89501-612EA 498.88 USD InStock
89501-612 CA48219-492
Extrelut Nt Refill Pack For Column Fillings Extrelut Nt 20
Chromatography Columns
EXtrelut® NT is a specially processed, inert, wide-pore kieselguhr with a high pore volume.
  • Saves solvent
  • Easy to use
  • Highly efficient
Other applications include environmental and residue testing such as analysis of industrial, domestic and waste water. EXtrelut® NT is supplied either pre-packed in cartridges or as a loose sorbent. EXtrelut® NT simplifies liquid-liquid extraction by replacing separation funnels. Using a single step is more efficient, saves solvent, as well as material and time, in contrast to classical funnel separation. The aqueous sample is simply applied to the EXtrelut® NT sorbent. It distributes itself over the chemically inert matrix and thus acts as a stationary phase. Subsequently, elution takes place using organic solvents that are non miscible with water. All the lipophilic substances are extracted from the aqueous into the organic phase.
It has been used for quite some time within research, for the sample preparation of urine, whole blood, plasma and other biological matrices.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty