Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Quinic acid for synthesis, for resolution of racemates, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

M-Clarity™ quality level = MQ200

8.00218.0050 8.00218.0050
EM8.00218.0050CS 19695 USD InStock
EM8.00218.0050 CA8.00218.0050
D-(-)-Quinic acid for synthesis, for resolution of racemates, Sigma-Aldrich®
D(-)-Quinic acid


Formula: C₇H₁₂O₆
MW: 192.17 g/mol
MDL Number: MFCD00003864
CAS Number: 77-95-2
EINECS: 201-072-8
Order Now
Specification Test Results
Solubility 290 g/l (9 °C) (experimental)
Melting Point 162.5 °C
Molar Mass 192.17 g/mol
Bulk Density 670 - 750 kg/m3
Density 1.64 g/cm3 (20 °C)
pH 2.0 (10 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty