Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dimethylurea for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: N,N'-Dimethylurea

M-Clarity™ quality level = MQ200

8.03244.1000 8.03244.1000
EM8.03244.1000CS 565 USD InStock
CA8.03244.1000 EM8.03244.1000
1,3-Dimethylurea for synthesis, Sigma-Aldrich®
1,3-Dimethylurea


Formula: C₃H₈N₂O
MW: 88.11 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 96 °C
Density: 1.142 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008286
CAS Number: 96-31-1
EINECS: 202-498-7
Order Now
Specification Test Results
Solubility 765 g/l (21.5 °C)
Melting Point 105 - 108 °C
Molar Mass 88.11 g/mol
Bulk Density 500 kg/m3
Boiling Point 268 - 270 °C (1013 hPa)
Vapor Pressure 6 hPa (115 °C)
Density 1.14 g/cm3 (20 °C)
pH 9.0 - 9.5 (100 g/l, H2O, 20 °C)
Ignition Point 400 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty