Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylacetonitrile for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2,2-Diphenylacetonitrile

M-Clarity™ quality level = MQ200

8.03259.0100 8.03259.0100
EM8.03259.0100CS 584 USD InStock
CA8.03259.0100 EM8.03259.0100
Diphenylacetonitrile for synthesis, Sigma-Aldrich®
Diphenylacetonitrile


Formula: (C₆H₅)₂CHCN
MW: 193.25 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 71…74 °C
Density: 1.182 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001862
CAS Number: 86-29-3
EINECS: 201-662-5
Merck Index: 13,03348
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 71 - 74 °C
Molar Mass 193.24 g/mol
Boiling Point 175 - 177 °C (15 hPa)
Vapor Pressure 21.3 hPa (190 °C)
Flash Point 120 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty