Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionyl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.07063.0500 8.07063.0100
EM8.07063.0500CS 439 USD InStock
EM8.07063.0500 EM8.07063.0100
Propionyl chloride for synthesis, Sigma-Aldrich®
Propionyl chloride


Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 80 °C (1013 hPa)
Melting Pt: –94 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3 (8),II
Merck Index: 12,08013
Order Now
Specification Test Results
Solubility (20 °C) (rigorous decomposition)
Melting Point -94 °C
Molar Mass 92.52 g/mol
Boiling Point 80 °C (1013 hPa)
Vapor Pressure 106 hPa (20 °C)
Flash Point 6.5 °C
Refractive Index 1.4032 (20 °C, 589 nm)
Explosion Limit 3.6 - 11.9 %(V)
Density 1.06 g/cm3 (20 °C)
Ignition Point 270 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty