Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrimethoxysilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Trimethoxy(methyl)silane

M-Clarity™ quality level = MQ200

8.14178.0250 8.14178.0250
EM8.14178.0250CS 730 USD InStock
EM8.14178.0250 CA8.14178.0250
Methyltrimethoxysilane for synthesis, Sigma-Aldrich®
Methyltrimethoxysilane


Formula: C₄H₁₂O₃Si
MDL Number: MFCD00008342
CAS Number: 1185-55-3
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Molar Mass 136.22 g/mol
Boiling Point 102 - 103 °C (1013 hPa)
Flash Point 9 °C
Refractive Index 1.3646 (20 °C, 589 nm)
Density 0.96 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty