Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl trifluoromethanesulfonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.18035.0010 8.18035.0010
EM8.18035.0010CS 90916 USD InStock
EM8.18035.0010 CA8.18035.0010
Methyl trifluoromethanesulfonate for synthesis, Sigma-Aldrich®
Methyl trifluoromethanesulphonate


Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
MDL Number: MFCD00000409
CAS Number: 333-27-7
UN: 2920
ADR: 8 (3),II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Molar Mass 164.1 g/mol
Boiling Point 99 °C (1013 hPa)
Flash Point 38 °C
Refractive Index 1.3228 (25 °C, 589 nm)
Density 1.49 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty