Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azobenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.20118.0100 8.20118.0100 8.20118.0025 8.20118.0025
EM8.20118.0100CS 3282 USD InStock
CA8.20118.0025 CA8.20118.0100 EM8.20118.0100 EM8.20118.0025
Azobenzene for synthesis, Sigma-Aldrich®
Azobenzene


Formula: C₁₂H₁₀N₂
MW: 182.22 g/mol
MDL Number: MFCD00003022
CAS Number: 103-33-3
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Solubility 6.4 mg/l (25 °C)
Melting Point 66 - 69 °C
Molar Mass 182.23 g/mol
Boiling Point 291 - 294 °C (1013 hPa)
Vapor Pressure 0.00048 hPa (25 °C)
Flash Point 100 °C
Density 1.203 g/cm3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty