Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butylbenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: n-Butylbenzene

M-Clarity™ quality level = MQ200

8.20239.0100 8.20239.0100
EM8.20239.0100CS 1560 USD InStock
CA8.20239.0100 EM8.20239.0100
Butylbenzene for synthesis, Sigma-Aldrich®
Butylbenzene


Formula: C₁₀H₁₄
MW: 134.22 g/mol
MDL Number: MFCD00009463
CAS Number: 104-51-8
UN: 2709
ADR: 3,III
Order Now
Specification Test Results
Solubility 11.8 mg/l (25 °C)
Melting Point -88 °C
Molar Mass 134.22 g/mol
Boiling Point 183 °C
Vapor Pressure 1.33 hPa (23 °C)
Flash Point 58 °C
Refractive Index 1.4895 (20 °C, 589 nm)
Explosion Limit 0.8 - 5.8 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 410 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty