Would you like to log in again?
UV Mount Medium, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17996
100504-080EA 287.34 USD InStock
100504-080
UV Mount Medium, Electron Microscopy Science
Stains Microscopy and staining reagents
UV Mount Medium is a mounting medium that is designed to match the refractive index of JB-4 embedded sections
A mounting medium that is designed to match the refractive index of JB-4 embedded sections.
Cures within 2 minutes when exposed to long wave UV light(365nm) when applied to moist sections.
Sections are mounted after last staining water rinse.
Slides must be coverslipped.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty