Would you like to log in again?
Micro Stopcock, Glass Plug, Ace Glass
Supplier: Ace Glass
8139-10
89062-344EA 162.62 USD InStock
89062-344
Micro Stopcock, Glass Plug, Ace Glass
Stopcocks
Specifically designed for micro chemical apparatus
The solid glass plug and barrel are lapped together to give a precision fit suitable for micro chemical requirements. Plugs are interchangeable within the meaning of ASTM Standard E 675.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty