Would you like to log in again?
Lennox Broth, GrowCells
Supplier: GrowCells
MBLE-7062 MBLE-7060-12 MBLE-7062-6 MBLE-7060
101076-258CS 409.37 USD InStock
101076-258 101384-874 101076-260 101076-262
Lennox Broth, GrowCells
Media for Microbiology Prepared Media for Microbiology
Ready to use sterile bacterial culture media
Prepared with 18.2 megOhm water, 0.2 micron filtered and autoclaved in final packaging.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty