Would you like to log in again?
IntelliCAL™ Ammonia ISE Analysis Package, Hach
Supplier: Hach
ISENH3181AP ISENH3181AP
89501-114EA 1703.31 USD InStock
CA89501-114 89501-114
IntelliCAL™ Ammonia ISE Analysis Package, Hach
Electrodes ISE Electrodes
The IntelliCAL™ ammonia ISE analysis package contains the IntelliCAL™ ISENH3181 Ammonia ISE, 500 ml Ammonia electrode storage solution, 100 pack of Ammonia ISA powder pillows, 3 pack of ISENH3181 membrane modules, 500 ml of 1.0 mg/L nitrogen-ammonia standard solution, 500 ml of 10 mg/L nitrogen-ammonia standard solution and 500 ml of 100 mg/L nitrogen-ammonia standard solution.
  • Provides fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • User alerted when re-calibration is needed
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
IntelliCAL™ ISENH3181 is a digital, combination gas-sensing ammonia ion selective electrode (ISE) with a replaceable membrane module, refillable outer body, double junction reference and built-in temperature sensor. The IntelliCAL ISENH3181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENH3181 is ideal for measuring ammonia concentrations in wastewater, drinking water and general water quality applications.
Special features include easy-to-replace membrane modules. Integrated thermistor.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty