Would you like to log in again?
SoC Broth, Grow Cells
Supplier: GROWCELLS MS
MBLE-8003-4 MBLE-8003-48
100894-916PK 82.35 USD InStock
100894-916 100915-640
SoC Broth, Grow Cells
Media for Microbiology Prepared Media for Microbiology
Ready to use Super Optimal broth with Catabolite repression (SOC)
Prepared with 18.2 megOhm water, 0.45 micron sterile filtered and autoclaved in final packaging
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty