Would you like to log in again?
Baumgartner Needle Holder, Integra™ Miltex®
Supplier: Integra LifeSciences
8-40
11000-462EA 203.24 USD InStock
11000-462
Baumgartner Needle Holder, Integra™ Miltex®
Forceps
The Baumgartner Needle Holder has smoothly beveled box locks and jaw edges protect delicate suture material from snags or cutting
The jaw surface is milled to ensure a secure grasp and to prevent needle slippage or rotation
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty